EMAIL THIS PAGE TO A FRIEND
Organic letters
From small building blocks to complex molecular architecture.
PMID 16562893
Abstract
[reaction: see text] We describe a synthesis of a dendrimer-like amphiphile containing a flat rigid core and 12 hydrophobic and hydrophilic arms. We employ a modular approach based on stepwise protection chemistry starting from simple building blocks. The key feature of this approach is the absence of a polymerization step, which makes it applicable for linear monofunctionalized precursors of any kind. This strategy also allows for precise control of the number of arms and ensures their alternating arrangement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
225266
Biphenyl-4,4′-dicarboxylic acid, 97%
C14H10O4