EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Rhodium-catalyzed asymmetric addition of aryl- and alkenylboronic acids to isatins.
PMID 16596682
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
693537
R-MOP, ≥94%
C33H25OP
579386
trans-1-Heptenylboronic acid
C7H15BO2