EMAIL THIS PAGE TO A FRIEND
The Journal of steroid biochemistry and molecular biology
Systemic distribution and tissue localizations of human 17beta-hydroxysteroid dehydrogenase type 12.
PMID 16621523
Abstract
The 17beta-hydroxysteroid dehydrogenases (HSDs) are enzymes that catalyze the reduction of 17-ketosteroids or the oxidation of 17beta-hydroxysteroids. 17beta-HSD type 12, the most recently cloned member of this gene family, was classified into the 17beta-HSD family based on sequence homology, rather than steroid catalyzing activity. Meanwhile, it has been reported that 17beta-HSD type 12 may be involved in fatty acid synthesis. To better understand the role of 17beta-HSD type 12 in lipid metabolism, we determined the detailed systemic distribution and tissue localizations of 17beta-HSD type 12, which, due partly to the lack of antibodies, had not yet been studied. We carried out these investigations by quantitative reverse transcription (RT)-PCR, Northern blot analysis, and immunohistochemistry, using an antibody against 17beta-HSD type 12 that we have generated. 17beta-HSD type 12 is highly expressed in organs related to lipid metabolism such as liver, kidney, heart and skeletal muscle. 17beta-HSD type 12 is also detected in endocrine-related organs such as pancreas, pituitary gland, adrenal gland, testis and placenta, and in the gastrointestinal tract, which point to the possible involvement of 17beta-HSD type 12 in the regulation of lipid biosynthesis and steroid metabolism. These results support previous reports and solidify the possibility that 17beta-HSD type 12 may play critical roles in the physiological processes, such as fatty acid synthesis, in addition to the steroid metabolism.