EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The discovery of tetrahydrofluorenones as a new class of estrogen receptor beta-subtype selective ligands.
PMID 16632357
Abstract
Synthesis and derivatization of a series of substituted tetrahydrofluorenone analogs giving potent, ERbeta subtype selective ligands are described. Several analogs possessing ERbeta binding affinities comparable to 17beta-estradiol but with greater than 75-fold selectivity over ERalpha are reported.