EMAIL THIS PAGE TO A FRIEND
Best practice & research. Clinical anaesthesiology
Special cases: ketamine, nitrous oxide and xenon.
PMID 16634415
Abstract
Most general anaesthetic agents produce anaesthesia by increasing the activity of inhibitory gamma-aminobutyric acid type A receptors. The effects of ketamine, xenon and nitrous oxide on these receptors are, however, negligible. These anaesthetic agents potently inhibit excitatory N-methyl-D-aspartate receptors. Although these anaesthetic agents display some similar clinical features, such as potent analgesic effects, there are some important differences. Ketamine and nitrous oxide produce sympathomimetic effects, whereas xenon produces a sympatholytic effect. In addition, these anaesthetic agents return differential signals on clinical available anaesthetic depth monitors such as the bispectral index and mid-latency auditory evoked potential. Ketamine and nitrous oxide do not per se decrease the bispectral index. However, xenon decreases the bispectral index in a concentration-dependent manner. Similarly, ketamine and nitrous oxide do not suppress the mid-latency auditory evoked potential whereas xenon does. Thus, anaesthetic depth monitors fail to describe consciousness accurately when ketamine and nitrous oxide are used.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00472
Xenon, ≥99.995%
Xe