EMAIL THIS PAGE TO A FRIEND
Expert opinion on pharmacotherapy
Chemoprevention of prostate cancer with finasteride.
PMID 16634712
Abstract
Prostate cancer is a significant cause of disease and death, making it an attractive target for chemoprevention. The association between lifetime exposure to dihydrotestosterone and risk of developing prostate cancer suggests that chemoprevention is possible with 5alpha-reductase inhibition. The recently completed Prostate Cancer Prevention Trial indicates that chemoprevention is possible with the 5alpha-reductase inhibitor finasteride. Development of a cost-effective chemoprevention strategy for prostate cancer is evolving, and is expected to have significant positive economic and public health benefits.