EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Dipeptidyl nitriles as human dipeptidyl peptidase I inhibitors.
PMID 16647256
Abstract
Using a dipeptide nitrile scaffold we have identified a potent and selective inhibitor of human dipeptidyl peptidase I.