EMAIL THIS PAGE TO A FRIEND
Biochemistry
Mechanism of inactivation of gamma-cystathionase by the acetylenic substrate analogue propargylglycine.
PMID 16648
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
81838
L-C-Propargylglycine, ≥99.0% (TLC)
C5H7NO2