EMAIL THIS PAGE TO A FRIEND
Organic letters
Rearrangement of homoallylic alcohols induced by DAST.
PMID 16671789
Abstract
[reaction: see text] Treatment of beta,gamma-unsaturated monoprotected 1,2-diols with diethylaminosulfur trifluoride (DAST) allows the stereoselective formation of beta,gamma-unsaturated aldehydes in good yields and with a good transfer of chirality.
Related Materials