EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Identification of enzyme responsible for erythritol utilization and reaction product in yeast Lipomyces starkeyi.
PMID 16716937
Abstract
We have identified the enzyme responsible for erythritol utilization and its reaction product in the yeast Lipomyces starkeyi CBS 1807. The enzyme, a polyol dehydrogenase requiring NAD+ as a coenzyme, was induced by erythritol in this yeast. We confirmed that the enzyme product was L-erythrulose by MS, NMR, and polarimeter analyses, meaning that we clarified the first step of erythritol utilization in yeasts for the first time. In the case of the oxidative reaction, D-threitol, (2R,3R)-2,3-butanediol, and erythritol were much better substrates than 21 other polyols tested. These three substrates are tetroses and have an R configuration at C-3, and whose third carbon results in easiest oxidation in this enzyme. The research of the substrate specificity in the reductive reaction demonstrated that L-erythrulose and dihydroxyacetone were better substrates, that D-acetoin was inactive and L-erythrose (aldose) was slightly active.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0758
L(+)-Erythrose, ~40%, syrup
C4H8O4