EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The synthesis and in vitro testing of structurally novel antibiotics derived from acylnitroso Diels-Alder adducts.
PMID 16730172
Abstract
The structural similarity between beta-lactam antibiotics, such as penicillin, and isoxazolidine-3,5-dicarboxylic acids led to the hypothesis that isoxazolidine-3,5-dicarboxylic acids could be effective analogs of beta-lactam antibiotics. The syntheses of relevant isoxazolidine-3,5-dicarboxylic acids from acylnitroso Diels-Alder adducts and subsequent biological testing have shown that these first examples are inhibitors of Escherichia coli X580.