EMAIL THIS PAGE TO A FRIEND
Veterinary immunology and immunopathology
Leukocyte antigens in cattle, sheep and goats. Proceedings of the First International Workshop on Leukocyte Antigens in Cattle, Sheep and Goats. Hannover, Germany, 25 July 1989.
PMID 1673576