EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Association of Catsper1 or -2 with Ca(v)3.3 leads to suppression of T-type calcium channel activity.
PMID 16740636
Abstract
Sperm-specific CatSper1 and CatSper2 proteins are critical to sperm-hyperactivated motility and male fertility. Although architecturally resembling voltage-gated ion channels, neither CatSper1 nor CatSper2 alone forms functional ion channels in heterologous expression systems, which may be related to the absence of yet unidentified accessory subunits. Here we isolated CatSper1- and CatSper2-associated protein(s) from human sperm and analyzed their identities by a multidimensional protein identification technology approach. We identified the T-type voltage-gated calcium channel Ca(v)3.3 as binding to both CatSper1 and CatSper2. The specificity of their interactions was verified by co-immunoprecipitation in transfected mammalian cells. Electrophysiological studies revealed that the co-expression of CatSper1 or CatSper2 specifically inhibited the amplitude of Ca(v)3.3-evoked T-type calcium current without altering other biophysical properties of Ca(v)3.3. Immunostaining studies revealed co-localization of CatSper1 and Ca(v)3.3 on the principal piece of human sperm tail. Furthermore, fluorescence resonance energy transfer analysis revealed close proximity and physical association of these two proteins on the sperm tail. These studies demonstrate that CatSper1 and CatSper2 can associate with and modulate the function of the Ca(v)3.3 channel, which might be important in the regulation of sperm function.