EMAIL THIS PAGE TO A FRIEND
Gastroenterology
HCCR-1-interacting molecule "deleted in polyposis 1" plays a tumor-suppressor role in colon carcinogenesis.
PMID 16762630
Abstract
Human cervical cancer oncogene (HCCR-1) has appeared to act as a negative regulator of p53 and contributes to tumorigenesis of various organs including the colon. We identified the HCCR-1 binding protein deleted in polyposis 1 (DP1) and accessed the role of HCCR-1 and DP1 in colon tumorigenesis. Yeast 2-hybrid was used to identify HCCR-1 interacting proteins. Various molecular biological approaches were used to examine the expression profile of HCCR-1 and DP1, subcellular localization, epitope mapping, the biological role of DP1, and the serum HCCR-1 level. Loss of heterozygosity frequency around DP1 also was examined. We identified that HCCR-1 interacted with DP1. These 2 proteins colocalized in mitochondria but the expression of HCCR-1 showed negative correlation with that of DP1 in colorectal cancer (CRC). DP1 played a tumor-suppressor role in colon tumorigenesis (ie, DP1-transfected RKO cells showed growth inhibition, apoptosis, decreased telomerase activity, and up-regulation of p53). These phenomena were reversed when HCCR-1 was overexpressed. Loss of heterozygosity around the DP1 gene was observed frequently (50%) in CRCs. We examined the use of serum HCCR-1 in CRC patients. The sensitivity of HCCR-1 (76.0%) for detecting CRC was proven to be much higher than that of CA19-9 (32.0%). DP1 plays a tumor-suppressor role in CRC. DP1 and HCCR-1 are supposed to regulate each other negatively by interaction, but further study is required to get better insight into the biological significance of the interaction.