EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
DyI2 initiated mild and highly selective silyl radical-catalyzed cyclotrimerization of terminal alkynes and polymerization of MMA.
PMID 16767277
Abstract
An efficient method for the formation of SiCl3 radicals by the reaction of abundant and cheaper chlorosilanes with DyI2 has been established, not only demonstrating new distinctive reactivities of solvated DyI2 but also suggesting that the presence of lanthanide ions can improve the selectivity of some silyl radical-catalyzed reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
652423
Dysprosium(II) iodide, anhydrous, powder, ≥99.9% trace metals basis
DyI2