EMAIL THIS PAGE TO A FRIEND
Journal of obstetrics and gynaecology Canada : JOGC = Journal d'obstetrique et gynecologie du Canada : JOGC
Canadian contraception consensus--update on Depot Medroxyprogesterone Acetate (dmpa).
PMID 16776909