EMAIL THIS PAGE TO A FRIEND
Cancer research
Antibodies to proliferating cell nuclear antigen as S-phase probes in flow cytometric cell cycle analysis.
PMID 1678682
Abstract
The usefulness of different anti-proliferating cell nuclear antigen monoclonal antibodies as S-phase probes in flow cytometric analysis was evaluated after various fixation procedures on human cell lines. With a newly developed detergent extraction/fixation method the monoclonal antibody PC10 acted as a selective S-phase marker. A total of 27 human hematopoietic tumors were analyzed using PC10, and all exhibited the characteristic S-phase recognition pattern. The percentage of PC10-positive cells was easily calculated and showed strong correlation to the fraction of S-phase cells determined from DNA histograms. Using alternative fixation procedures PC10, on the other hand, could recognize all actively cycling cells, a feature also observed for the monoclonal antibodies, 19A2 and TOB7.