EMAIL THIS PAGE TO A FRIEND
Tetrahedron, asymmetry
Enantioselective radical reactions. Evaluation of nitrogen protecting groups in the synthesis of beta-amino acids.
PMID 16799704
Abstract
We have investigated the effect of nitrogen protecting groups in radical addition trapping experiments leading to beta(2)-amino acids. Of the three N-protecting groups examined, the phthalimido group was optimal with respect to both yields and enantioselectivity. Additionally, radical additions to more complex acrylates were also investigated, which provided access to functionalized beta(2)-amino acids in modest selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
302546
Methyl 2-(bromomethyl)acrylate, 97%
C5H7BrO2