EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Nondestructive amino acid analysis at the picomole level of proline-containing peptides using aminopeptidase M and prolidase: application to peptides containing tyrosine sulfate.
PMID 1683181
Abstract
A micromethod for nondestructive amino acid analysis was developed. Peptides were digested with amino-peptidase M followed by prolidase. The whole procedure was performed in a microvial for the autosampler of the analyzer. The resulting method has a working range from 10 to 600 pmol, at least, of sample taken into work. The usefulness of the method was demonstrated by its ability to determine the sulfation of tyrosine preceding proline in chicken gastrin.