EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Online synthesis of diblock cationic oligonucleotides for enhanced hybridization to their complementary sequence.
PMID 16874748
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
512028
tert-Butyl(4-iodobutoxy)dimethylsilane, 95%
C10H23IOSi