EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Polynucleotide kinase exchange reaction: quantitave assay for restriction endonuclease-generated 5'-phosphoroyl termini in DNA.
PMID 16886