EMAIL THIS PAGE TO A FRIEND
Zhongguo yi xue ke xue yuan xue bao. Acta Academiae Medicinae Sinicae
[Itraconazole in the treatment of superficial candidal infections: twelve years' clinical experience].
PMID 16900653
Abstract
Itraconazole has been used to treat superficial candidal infections in China for 12 years with promising efficacy and safety. This article retrospectively reviewed literatures published in the mainstream journals in China with an attempt to find a reasonable therapy for Chinese populations.