EMAIL THIS PAGE TO A FRIEND
Organic letters
Generation of aza-ortho-xylylenes via ring opening of 2-(2-acylaminophenyl)aziridines: application in the construction of the communesin ring system.
PMID 16928057
Abstract
A new protocol for generating aza-ortho-xylylenes via acid-catalyzed or fluoride-promoted ring opening of 2-(2-acylaminophenyl)aziridines is described. This methodology has been exploited in the rapid construction of a hexacyclic substructure of communesin B.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226890
2-(Trimethylsilyl)ethanol, 99%
C5H14OSi