EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
One-step purification of trypsin and alpha-chymotrypsin by affinity chromatography on Eupergit-aprotinin, a novel carrier for purification of serine proteases.
PMID 1698197
Abstract
Aprotinin isolated from bovine lungs was covalently immobilized on Eupergit C. The highly selective affinity adsorbent was used to purify trypsin and alpha-chymotrypsin from pancreatic extract in a single step. Sodium dodecyl sulphate-polyacrylamide gel electrophoresis indicated that both enzymes were highly purified. The maximum binding capacity of Eupergit-aprotinin for bovine trypsin was calculated to be ca. 7.5 mg per gram of gel (wet resin).