EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Rhenium-catalyzed insertion of aldehyde into a C-H bond: synthesis of isobenzofuran derivatives.
PMID 16984162
Abstract
A rhenium complex, [ReBr(CO)3(thf)]2, catalyzed reactions of aromatic ketimines with aldehydes to give isobenzofuran derivatives in good to excellent yields. In contrast to ruthenium and rhodium catalysts, aldehydes, which are polar unsaturated molecules, inserted into the C-H bond after activation by the rhenium complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
685615
Bromotricarbonyl(tetrahydrofuran)rhenium(I) dimer
C14H16Br2O8Re2