EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Metal complexes with superoxide dismutase-like activity as candidates for anti-prion drug.
PMID 16987659
Abstract
Various compounds were evaluated for ability to inhibit the formation of the abnormal protease-resistant form of prion protein (PrP-res) in two cell lines infected with different prion strains. Examination of the structure-activity relationships indicated that compounds with copper-selective chelating ability and whose copper complexes have high SOD-like activity are candidates for anti-prion drug.