EMAIL THIS PAGE TO A FRIEND
Nature structural & molecular biology
Structural analysis of kasugamycin inhibition of translation.
PMID 16998486
Abstract
The prokaryotic ribosome is an important target of antibiotic action. We determined the X-ray structure of the aminoglycoside kasugamycin (Ksg) in complex with the Escherichia coli 70S ribosome at 3.5-A resolution. The structure reveals that the drug binds within the messenger RNA channel of the 30S subunit between the universally conserved G926 and A794 nucleotides in 16S ribosomal RNA, which are sites of Ksg resistance. To our surprise, Ksg resistance mutations do not inhibit binding of the drug to the ribosome. The present structural and biochemical results indicate that inhibition by Ksg and Ksg resistance are closely linked to the structure of the mRNA at the junction of the peptidyl-tRNA and exit-tRNA sites (P and E sites).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K4013
Kasugamycin hydrochloride from Streptomyces kasugaensis, ≥90% (HPLC)
C14H25N3O9 · HCl