EMAIL THIS PAGE TO A FRIEND
Organic letters
Radical trifluoromethylation of ketone silyl enol ethers by activation with dialkylzinc.
PMID 17020274
Abstract
[reaction: see text] The radical trifluoromethylation of ketone silyl enol ethers gave alpha-CF(3) ketones in good yields with wide scope of the ketonic substrates including acyclic ketones and cyclopentanone. The use of dialkylzinc to activate the silyl enol ethers is the key to the efficient radical trifluoromethylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
364401
Dimethylzinc solution, 2.0 M in toluene
C2H6Zn