EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Antitumor studies. Part 1: design, synthesis, antitumor activity, and AutoDock study of 2-deoxo-2-phenyl-5-deazaflavins and 2-deoxo-2-phenylflavin-5-oxides as a new class of antitumor agents.
PMID 17049252
Abstract
Novel 2-deoxo-2-phenyl-5-deazaflavins and 2-deoxo-2-phenylflavin-5-oxides were prepared as a new class of antitumor agents and showed significant antitumor activities against NCI-H 460, HCT 116, A 431, CCRF-HSB-2, andKB cell lines. In vivo investigation, 2-deoxo-10-methyl-2-phenyl-5-deazaflavin exhibited the effective antitumor activity against A 431 human adenocarcinoma cells transplanted subcutaneously into nude mouse. Furthermore, AutoDock study has been done by binding of the flavin analogs into PTK pp60(c-src), where a good correlation between their IC(50) and AutoDock binding free energy was exhibited. In particular, 2-deoxo-2-phenylflavin-5-oxides exhibited the highest potential binding affinity within the binding pocket of PTK.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1768
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
C9H13N3O5