EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
BF3.OEt2 and TMSOTf: A synergistic combination of Lewis acids.
PMID 17057868
Abstract
The combination of BF3.OEt2 and TMSOTf gives BF2OTf.OEt2, which is a more powerful Lewis acid than its components and especially effective in CH3CN solvent; the complex formed has been characterised by 1H, 19F, 11B and 31P (using Et3PO as an additive) NMR spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
225649
Trimethylsilyl trifluoromethanesulfonate, 99%
C4H9F3O3SSi