EMAIL THIS PAGE TO A FRIEND
Organic letters
Silylative Dieckmann-like cyclizations of ester-imides (and diesters).
PMID 17078675
Abstract
[Structure: see text] Trialkylsilyl triflates effect cyclization of ester-imides such as 2 to produce adducts such as 4a. Trapping of the in situ generated, nucleophilic ketene acetal (cf. 5a) is a key aspect of the transformation. A range of substrates amenable to this operationally simple reaction is reported. In many instances the levels of diastereoselectivity are very high. Mechanistic points are inferred from spectroscopic observations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
225649
Trimethylsilyl trifluoromethanesulfonate, 99%
C4H9F3O3SSi