EMAIL THIS PAGE TO A FRIEND
Cell
Global, in vivo, and site-specific phosphorylation dynamics in signaling networks.
PMID 17081983
Abstract
Cell signaling mechanisms often transmit information via posttranslational protein modifications, most importantly reversible protein phosphorylation. Here we develop and apply a general mass spectrometric technology for identification and quantitation of phosphorylation sites as a function of stimulus, time, and subcellular location. We have detected 6,600 phosphorylation sites on 2,244 proteins and have determined their temporal dynamics after stimulating HeLa cells with epidermal growth factor (EGF) and recorded them in the Phosida database. Fourteen percent of phosphorylation sites are modulated at least 2-fold by EGF, and these were classified by their temporal profiles. Surprisingly, a majority of proteins contain multiple phosphorylation sites showing different kinetics, suggesting that they serve as platforms for integrating signals. In addition to protein kinase cascades, the targets of reversible phosphorylation include ubiquitin ligases, guanine nucleotide exchange factors, and at least 46 different transcriptional regulators. The dynamic phosphoproteome provides a missing link in a global, integrative view of cellular regulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7634 3-Phosphoglyceric Phosphokinase from baker's yeast (S. cerevisiae), ammonium sulfate suspension, ≥1000 units/mg protein
A6986 Acetyl-CoA Carboxylase 1 human, recombinant, expressed in Sf9 cells
A6861 Acetyl-CoA carboxylase 2 human, recombinant, expressed in Sf9 cells
A7510 Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
A8811 Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein
A2714
Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein
A9329 Aldolase from spinach, lyophilized powder, 0.3-1.0 units/mg protein
G5262 GAPDH, standard for protein electrophoresis
G0389 Glyceraldehyde-3-phosphate Dehydrogenase Agarose from baker's yeast (S. cerevisiae), lyophilized powder, 50-150 units/g dry agarose, pH 7.6, 30 °C
G5537 Glyceraldehyde-3-phosphate Dehydrogenase from baker's yeast (S. cerevisiae), lyophilized powder, 70-140 units/mg protein
G9263
Glyceraldehyde-3-phosphate Dehydrogenase from chicken muscle, lyophilized powder, ≥40 units/mg protein
G6019 Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes, lyophilized powder, 50-150 units/mg protein
G2267
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
I1782 Inosine Monophosphate Dehydrogenase Type II human, recombinant, expressed in E. coli
MSP1H MS PhosphoMix 1 Heavy, Phosphopeptide Standard for MS
MSP1L MS PhosphoMix 1 Light, Phosphopeptide Standard for MS
MSP2H MS PhosphoMix 2 Heavy, Phosphopeptide Standard for MS
MSP2L MS PhosphoMix 2 Light, Phosphopeptide Standard for MS
MSP3H MS PhosphoMix 3 Heavy, Phosphopeptide Standard for MS
MSP3L MS PhosphoMix 3 Light, Phosphopeptide Standard for MS
P3397
Phosphoglucomutase from rabbit muscle, ammonium sulfate suspension, ≥100 units/mg protein
P8913
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein
P5910 Phospholipase A2 from Crotalus durissus terrificus venom, lyophilized powder, ~200 units/mg protein
P9279 Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
P7778 Phospholipase A2 from Naja mossambica mossambica, lyophilized powder, ~1,500 units/mg protein
P6534 Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
P8685 Phospholipase A2 from Streptomyces violaceoruber, lyophilized powder, ≥10 units/mg solid
P0515 Phospholipase D from Arachis hypogaea (peanut), Type II, lyophilized powder, ≥60 units/mg protein
P8398
Phospholipase D from cabbage, Type IV, lyophilized powder, ≥100 units/mg solid
P0065 Phospholipase D from Streptomyces chromofuscus, ≥50,000 units/mL, buffered aqueous glycerol solution