EMAIL THIS PAGE TO A FRIEND
Veterinary immunology and immunopathology
Individual antigens of cattle. Bovine CD6 (BoCD6).
PMID 1708556
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91072526
IL-A57, Reactivity: Anti-CD6, bovine