EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Inhibition of CBF/NF-Y mediated transcription activation arrests cells at G2/M phase and suppresses expression of genes activated at G2/M phase of the cell cycle.
PMID 17098936
Abstract
Previous studies showed that binding of the CBF/NF-Y (CBF) transcription factor to cellular promoters is essential for cell proliferation. This observation prompted us to investigate the function of CBF in relation to cell cycle progression and in cell-cycle-regulated transcription. In this study, we used a tetracycline-inducible adenoviral vector to express a truncated CBF-B subunit, Bdbd, lacking a transcription activation domain in various mammalian cell lines. The Bdbd polypeptide interacts with cellular CBF-A/CBF-C and binds to promoters containing CBF-binding sites. Interestingly, expression of Bdbd in various mammalian cells resulted in the inhibition of cell proliferation and specific cell cycle arrest at G2/M phase. Gene expression analysis demonstrated that the expression of Bdbd strongly suppressed cell cycle-dependent transcription activation of Cyclin B1, Aurora A and CDK1 genes, key regulators for cell cycle progression at G2/M phase. Chromatin immunoprecipitation analysis showed that Bdbd significantly inhibited binding of TATA-binding protein, TBP to both Cyclin B1 and Aurora A promoters, but did not inhibit binding of E2F3 activator to Cyclin B1 promoter. This study suggested that the activation domain of CBF-B plays an essential role in the transcription activation of Cyclin B1 and Aurora A genes at G2/M phase, thus regulating cell cycle progression at G2/M phase.