EMAIL THIS PAGE TO A FRIEND
Organic letters
Cobalt-catalyzed aryl-sulfur bond formation.
PMID 17107085
Abstract
A new cobalt-catalyzed coupling of aryl halides with thiophenols and alkanethiols is reported. A variety of aryl sulfides can be prepared in excellent yields under mild reaction conditions using 1-2 mol % of CoI2(dppe) and Zn. This new cobalt-catalyzed coupling represents an interesting addition to previously known methods to synthesize thioethers. [reaction: see text].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
206555
4-Iodotoluene, 99%
C7H7I
685631
Diiodo(bis(diphenylphosphino)ethane)cobalt(II)
C26H24CoI2P2