EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Highly activated vinyl hydrogen in a significantly twisted styrene.
PMID 17109523
Abstract
The novel example of a vinylic hydrogen more reactive than a benzylic hydrogen was found by treatment of a twisted styrene derivative with a strong base followed by D(2)O quenching. In this paper, the full details of the examples of the highly activated vinyl hydrogens in twisted styrene derivatives are described, with a discussion on the correlation between the reactivity of the vinyl hydrogens and the magnitude of the twist. The highly reactive vinyl hydrogens could be rationalized by considering the novel orbital interaction between the pi(*) orbital of the benzene ring and the pi(*) orbital of the vinylic C-H bond in the twisted styrene derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
411019
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
C6H16N2