EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
CARPs are ubiquitin ligases that promote MDM2-independent p53 and phospho-p53ser20 degradation.
PMID 17121812
Abstract
Caspase 8/10-associated RING proteins (CARPs) are a recently described family of protein ubiquitin ligases that interact with and negatively regulate death receptor-mediated apoptosis. Because CARPs are overexpressed in cancer and their silencing reduces cell viability and sensitizes tumor cells to chemotherapeutic agents, we investigated their relationship to p53 tumor suppressor signaling. p53 is a major determinant of chemosensitivity, and its levels are increased following DNA damage through N-terminal phosphorylation and inhibition of degradation. Although p53 is well known to be negatively regulated by several ubiquitin ligases including MDM2, none are known to target phosphorylated p53 for degradation. CARPs physically interact with and ubiquitinate p53, targeting it for degradation in the absence of MDM2. Serine 20-phosphorylated p53 is also ubiquitinated by CARPs. CARP silencing stimulates p53 expression and promotes downstream effects, including transcriptional activation and tumor suppression.