EMAIL THIS PAGE TO A FRIEND
The American journal of surgical pathology
Glypican 3: a novel marker in testicular germ cell tumors.
PMID 17122513
Abstract
Glypican 3 (GPC3), a membrane-bound heparin sulfate proteoglycan, may play a role in promoting embryonic cell growth and differentiation. GPC3 is mutated in Simpson-Golabi-Behmel syndrome, characterized by tissue overgrowth and an increased risk of embryonal malignancies. Recently, GPC3 was reported to be one of the over-expressed genes in testicular yolk sac tumors by gene expression microarray analysis. However, the presence of the GPC3 protein in germ cell tumors has never been investigated. The purpose of the study was to investigate the GPC3 expression in various histologic components of testicular germ cell tumors using immunohistochemistry and to assess its possible utility as a diagnostic marker. Tumors from 71 patients were examined, including components of 42 seminomas, 37 embryonal carcinomas, 24 yolk sac tumors, 20 teratomas with mature elements, 16 teratomas with immature elements, and 7 choriocarcinomas. Cytoplasmic and membranous immunoreactivity was semiquantitatively evaluated. All yolk sac tumor (24/24) and choriocarcinoma (7/7) components were immunoreactive for GPC3, whereas only 38% of teratomas with immature elements and 8% of embryonal carcinomas expressed GPC3. There was no immunoreactivity in benign testicular tissue, intratubular germ cell neoplasia, seminomas (0/42), or teratomas with mature elements (0/20). We conclude that the oncofetal protein GPC3 is a novel immunohistochemical marker in testicular germ cell tumors with differential expression in defined histologic subtypes. Our findings suggest a possible role of GPC3 in tumor cell differentiation. Furthermore, GPC immunostaining may be useful in the pathologic diagnosis of nonseminomatous germ cell tumors, particularly yolk sac tumor, and choriocarcinoma.