EMAIL THIS PAGE TO A FRIEND
Current opinion in urology
Holmium laser treatment of benign prostatic hyperplasia: an update.
PMID 17143108
Abstract
The surgical treatment of benign prostatic hyperplasia is a dynamic, evolving field. Holmium laser enucleation of the prostate has been one of the most rigorously analyzed interventions for benign prostatic hyperplasia. In the 12 months since July 2005, a number of important studies have been published concerning this technique. In the 12-month period of this review, there have been a number of articles published on holmium laser enucleation of the prostate. Among these are five randomized controlled trials. These studies emphasize the unique advantages of holmium laser enucleation of the prostate over other surgical treatments for benign prostatic hyperplasia. Holmium laser enucleation of the prostate is a more efficient procedure than competitor techniques, when grams of tissue removed per unit time are quantified. Additionally, holmium laser enucleation of the prostate is associated with a reduced length of catheterization and hospitalization when compared with other surgical therapies for men with benign prostatic hyperplasia. Outcome measures for men undergoing holmium laser enucleation of the prostate are in many cases superior to those of other modalities. It is likely that the completeness of adenoma removal with holmium laser enucleation of the prostate confers many of these advantages.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
457957
Holmium, chips, 99.9% trace metals basis
Ho
693693
Holmium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Ho
755974
Holmium, dendritic pieces, purified by distillation, 99.99% trace rare earth metals basis
Ho
GF92551259
Holmium, rod, 50mm, diameter 3.8mm, 99%
Ho
GF12230154
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Ho
GF39809863
Holmium, wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF22997188
Holmium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ho
GF59427513
Holmium, wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF74648223
Holmium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Ho
GF88732693
Holmium, wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF13376785
Holmium, rod, 200mm, diameter 3.8mm, 99%
Ho
GF28769541
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Ho
GF68907011
Holmium, rod, 100mm, diameter 3.8mm, 99%
Ho
GF31028543
Holmium, powder, 5g, max. particle size 500 micron, 99.9%
Ho
GF78567654
Holmium, powder, 10g, max. particle size 500 micron, 99.9%
Ho
GF29991673
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ho
GF26625067
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Ho
GF42613006
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Ho
GF60938635
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Ho
GF34764666
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Ho
GF78609640
Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Ho
GF49265253
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Ho
GF07383977
Holmium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ho
GF85502640
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Ho
GF19613684
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Ho
GF38519376
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Ho