EMAIL THIS PAGE TO A FRIEND
The Medical letter on drugs and therapeutics
Nabilone (Cesamet) for chemotherapy-induced nausea and vomiting.
PMID 17149361
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3785
Nabilone, solid, ≥98% (HPLC)
C24H36O3