EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Inhibitors of the FEZ-1 metallo-beta-lactamase.
PMID 17157014
Abstract
Metallo-beta-lactamases (MBLs) catalyze the hydrolysis of beta-lactams including penicillins, cephalosporins and carbapenems. Starting from benzohydroxamic acid (1) structure-activity studies led to the identification of selective inhibitors of the FEZ-1 MBL, e.g., 2,5-substituted benzophenone hydroxamic acid 17 has a K(i) of 6.1+/-0.7microM against the FEZ-1 MBL but does not significantly inhibit the IMP-1, BcII, CphA or L1 MBLs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
412260
Benzhydroxamic acid, 99%
C7H7NO2