EMAIL THIS PAGE TO A FRIEND
Current opinion in chemical biology
Chemical genetic approaches to probing cell death.
PMID 17174591
Abstract
Chemical genetics has arisen as a tool for the discovery of pathways and proteins in mammalian systems. This approach, comprising small-molecule screening combined with biochemical and genomic target identification methods, enables one to assess which proteins are involved in regulating a particular phenotype. Applied to cell death, this strategy can reveal novel targets and pathways regulating the demise of mammalian cells. Numerous diseases have been linked to the loss of regulation of cell death. Defining the mechanisms governing cell death in these diseases might lead to the discovery of therapeutic agents and targets and provide a richer understanding of the mortality of living systems. Recent advances include the discovery of novel small molecules regulating cell death pathways -- necrostatin and erastin -- as well as the elucidation of the mechanism of death induced in cancer cells by the cytotoxic agent Apratoxin A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7781
Erastin, ≥98% (HPLC)
C30H31ClN4O4