EMAIL THIS PAGE TO A FRIEND
Drugs
Famciclovir: a review of its use in herpes zoster and genital and orolabial herpes.
PMID 17181386
Abstract
Famciclovir (Famvir) is the oral prodrug of penciclovir, an agent that has demonstrated antiviral activity against herpes simplex viruses, type 1 (HSV-1) and 2 (HSV-2) [which cause orolabial and/or genital herpes simplex], and against varicella zoster virus (VZV) [a reactivation of which leads to herpes zoster]. Famciclovir has efficacy similar to that of aciclovir (in immunocompetent or immunocompromised patients) or valaciclovir (in immunocompetent patients) in the treatment of herpes zoster, and efficacy similar to aciclovir in the treatment of first or recurrent episodes of genital herpes (in immunocompetent or immunocompromised patients). Famciclovir also has efficacy in the suppression of recurrent episodes of genital herpes, and in the treatment of orolabial herpes, in immunocompetent patients. As such, famciclovir is a well tolerated first-line option for the treatment of herpes zoster and the treatment and suppression of genital herpes, and is approved for the treatment of recurrent orolabial herpes. Convenient patient-initiated single-day (for recurrent genital herpes) and single-dose (for orolabial herpes) dosage regimens may contribute to treatment compliance, patient acceptability and subsequent treatment outcomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7932
Famciclovir, ≥98% (HPLC)
C14H19N5O4