EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Ternary nickel(II) complexes as hydrolytic DNA-cleavage agents.
PMID 17193262
Abstract
A series of small model complexes made from Ni(II) and the ligands ethylenediamine (en), histamine (hist), and histidylleucine (HisLeu) were prepared and studied as potential hydrolytic DNA-cleavage agents. The stability constants and species-distribution curves for these complexes were determined as a function of pH. The 1 : 1 : 1 ternary complexes [Ni(II)(en)(HisLeu)] (1) and [Ni(II)(hist)(HisLeu)] (2) were the only major species present at the physiologically relevant pH of 6-7, as further corroborated by ESI-MS analysis. The complex geometries of 1 and 2 were analyzed by UV/VIS experiments and molecular dynamics (MD) simulations. Both ternary complexes were found to intercalate with DNA, as shown by UV/VIS, thermal-denaturation, and fluorescence-titration studies with ethidium bromide (EB). The intrinsic binding constants (K(b)) for the bound complexes 1DNA and 2DNA were determined as 150 and 290, resp. Gel-electrophoresis experiments revealed that 1 and 2 cleave supercoiled (type-I) to nicked-circular (type-II) DNA at physiological pH, with rate constants of 0.64 and 0.75 h(-1), resp. A tentative mechanism for this hydrolytic cleavage is proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2504
His-Leu
C12H20N4O3
H3129
His-Ser
C9H14N4O4