EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
N-Alkyl-N-alkyloxycarbonylaminomethyl (NANAOCAM) prodrugs of carboxylic acid containing drugs.
PMID 17196386
Abstract
Synthesis and hydrolysis in aqueous buffers of novel N-alkyl-N-alkyloxycarbonylaminomethyl (NANAOCAM) and N-aryl-N-alkyloxycarbonylaminomethyl (NArNAOCAM) derivatives of carboxylic acid containing drugs were carried out. The hydrolysis follows a S(N)1 type mechanism and is dependent on the nucleofugacity of the leaving group. Topical delivery of the NANAOCAM derivative of naproxen from IPM across hairless mice skin was examined in in vitro diffusion cell experiments. The prodrug was 4.5-fold less lipid soluble, 2.4-fold less water soluble and 3.6-fold less permeable than the parent drug.