EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Combination of tigecycline and N-acetylcysteine reduces biofilm-embedded bacteria on vascular catheters.
PMID 17220399
Abstract
To assess the efficacy of an antibiofilm/antimicrobial agent combination, we incubated catheter segments colonized with one of six studied bacterial organisms in N-acetylcysteine, tigecycline, N-acetylcysteine-tigecycline, or saline. Segments were washed, sonicated, and cultured. N-acetylcysteine-tigecycline significantly decreased all viable biofilm-associated bacteria and was synergistic for methicillin-resistant Staphylococcus aureus and Staphylococcus epidermidis.