EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
New thiopyrazolo[3,4-d]pyrimidine derivatives as anti-mycobacterial agents.
PMID 17239593
Abstract
The multiple parallel synthesis of a series of N,S-bis-alkylated thiopyrazolo[3,4-d]pyrimidines, based on sequential S- then N-alkylation, is reported. These compounds showed significant anti-mycobacterial activity (MICs down to 2mug/ml) and their potential as significant drug-like leads is substantiated through cytotoxicity evaluation and in silico profiling.