EMAIL THIS PAGE TO A FRIEND
Helvetica chimica acta
[Hormone-receptor interactions. Syntheses of alpha-melanotropin and of informational sequences thereof with the aid of alcali-labile protecting groups (author's transl)].
PMID 172480