EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Facile heterolytic cleavage of dihydrogen by phosphines and boranes.
PMID 17260994
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442593
Tris(pentafluorophenyl)borane, 95%
C18BF15