EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Antistaphylococcal activity of DX-619 alone and in combination with vancomycin, teicoplanin, and linezolid assessed by time-kill synergy testing.
PMID 17261625
Abstract
Time-kill synergy studies testing in vitro activity of DX-619 alone and with added vancomycin, teicoplanin, or linezolid against 101 Staphylococcus aureus strains showed synergy between DX-619 and teicoplanin at 12 to 24 h in 72 strains and between DX-619 and vancomycin in 28 strains. No synergy was found with linezolid, and no antagonism was observed with any combination.